Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ValueWare® Reusable Soda Lime Glass Petri Dishes, DWK Life Sciences
Supplier: DWK Life Sciences
53061-10015 53061-6015 53061-10020 53061-10010
76175-260CS 319.54 USD InStock
76175-260 76175-266 76175-264 76175-262
ValueWare® Reusable Soda Lime Glass Petri Dishes, DWK Life Sciences
Culture Dishes Petri Dishes
These shallow glass dishes are manufactured from soda lime glass.
Sold in sets consisting of top and bottom dishes.
it is autoclavable at 121 °C and can withstand temperatures up to 500 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty