Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytofluid, Sterile, Quality Biological

Supplier: Quality Biological
114-205-161
76176-674EA 211.77 USD InStock
76176-674
Cytofluid, Sterile, Quality Biological
Cell Culture Supplements and Additives
Proprietary sheath fluid designed specifically for flow cytometry applications.

  • 1μm sterile filtered
  • Gamma irradiated to a minimum of 25kGy and maximum of 50kGy

Sterile cytofluid supports the applications of flow cytometry analyzation of cells, while reducing the probability of instrument contamination as opposed to non-sterile solutions. Cytofluid is 1X concentrated cube container with 20L sheath fluid for use in high-throughput fluidics (HTF) systems.

UNSPSC: 12161700
Harmonization Code: 3822.00.5090
ECCN: EAR99.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Delivery information: Ambient shipping conditions.

Caution: Research Use Only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty