Would you like to log in again?
FACEPIECE RU6500 SRS FULL NOSE CUP LARGE
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
RU6500 Series Full Facepiece Silicone Large^ RU6500 Series Full Facepiece, Silicone-Nose Cup, with mesh headnet-Nylon, North N-Series, seal is soft and flexible to provide workers with a comfortable fit, lightweight design, polycarbonate lens is designed to withstand high impact, Size: large
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACEPIECE RU6500 SRS FULL NOSE CUP LARGE 76176-724 RU65002L Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty