Would you like to log in again?
Histo-Tek® Mini Linear Slide Stainer, Sakura Finetek
Supplier: Sakura
1920
76176-726EA 9975.27 USD InStock
76176-726
Histo-Tek® Mini Linear Slide Stainer, Sakura Finetek
Slide Stainers
Automated staining for frozen section slides.
  • Simple to use keypad display
  • Continuous loading of up to 4 slides per carrier
  • 50 mL containers to minimize reagent waste and reduce costs
  • Fume cover and filter protects lab personnel
Staining time can be programmed from 2 seconds to 300 seconds for all stations. Program agitation from 0 to 3 dips per station, with intervals of 5 seconds. Start at any station, #1 to #14. Holds up to 16 slides and fits on any benchtop.
Certifications: IECO 61010 classification.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty