Would you like to log in again?
DNAdvance Kits, Beckman Coulter
Supplier: Beckman Coulter
DNAdvance
A48706 A48705 A48706 A48705
76177-125EA 1162.22 USD InStock
76177-125 CABKA48706 CABKA48705 76177-139
DNAdvance Kits, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Isolation from mammalian tissue. Built on SPRI paramagnetic bead-based technology, the DNAdvance Kit is a high throughput genomic DNA (gDNA) isolation reagent kit enabling the purification of high quality DNA from mammalian tissue samples.
  • Automated available
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty