Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ImageXpress® Micro Confocal High-Content Imaging System, Molecular Devices
Supplier: Molecular Devices
ImageXpress
IXMC-DB IXMC-DB
76177-140EA 454098.29 USD InStock
76177-140 CA76177-140
ImageXpress® Micro Confocal High-Content Imaging System, Molecular Devices
Photodocumentation Systems
Explore new dimensions with the confocal system for complex biology. The ImageXpress® Micro Confocal High-Content Imaging System enables users to expand the boundaries of research with the ultimate combination of speed, sensitivity, and flexibility in a turn-key solution.
  • Ideal for addressing the expanding need for three-dimensional (3D) models in higher throughput workflows
  • Improves mimic aspects of human tissues including the architecture, cell organization, cell-cell and cell-matrix interactions, and diffusion characteristics
  • Quickly and easily switch between a range of available confocal disk geometries and widefield imaging modes to satisfy the specific throughput and sensitivity needs for assays
  • Compatible with both acquisition and analysis of 2D and 3D cell models in classic screening, as well as generating 3D volumetric output and analysis for scale-up
The ImageXpress® Micro Confocal High-Content Imaging System helps expand the boundaries of research with the ultimate combination of speed, sensitivity, and flexibility
Capture high-quality images of whole organism, thick tissues, 3D spheroid assays, and cellular or intracellular events. Combined with MetaXpress® High-Content Image Acquisition and Analysis Software, the ImageXpress® Micro Confocal system provides a complete multi-dimensional, high-throughput screening solution to help discover the next landmark scientific breakthrough. Match acquisition mode to assay needs with multiple confocal geometries and widefield mode.
Quickly and easily switch between widefield and several confocal disk configurations, including an exclusive slit option, to capture high quality images at high-throughput speeds.
Capture publication-quality images without sacrificing throughput using the proprietary AgileOptix™ technology.
Proprietary AgileOptix™ technology provides increased sensitivity, resulting in high contrast and high uniformity of intensity and shape across a large field of view.
Acquire statistically relevant data faster with large field of view and >3 log dynamic range.
Featuring a high-quantum efficiency sCMOS detector and power-optimized solid-state light source, the ImageXpress® Micro Confocal system delivers images at the throughput of widefield systems.
High-content acquisition and image analysis software for complex biological assays.
MetaXpress® Software is optimized to perform with the ImageXpress® Micro imaging systems, allowing precise control over image acquisition and powerful, elegant tools for analysis. Gain speed and convenience with a single wizard guide through image acquisition and analysis, laser-based autofocus, pre-designed image analysis application modules, parallel processor support, and automatic data transfer scheduling. An interactive custom module editor transforms user knowledge, ideas, and intuition into entirely new, multiple-step analysis routines that can be created and run in minutes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty