Would you like to log in again?
Dispase (from Bacillus polymyxa)
Supplier: MP Biomedicals
0219502225 0219502205
76177-258EA 352.39 USD InStock
76177-258 76177-256
Dispase (from Bacillus polymyxa)
Enzymes
Dispase is an extremely stable microbial metalloprotease suitable for use in cell cultures.
  • Presentation: Tan Fluffy Powder
  • pH optimum: 6.0 - 8.5
Dispase is suitable for tissue disaggregation and subcultivation procedures since it does not damage cell membranes.
Dissolve the lyophilized enzyme in sterile water to 5 mg/mL. Further dilution with sterile PBS (Ca²⁺/Mg²⁺ free).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty