Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chicken Lysozyme Type VI (from Egg White), MP Biomedicals

Supplier: MP Biomedicals
0219530301
76177-422EA 81.03 USD InStock
76177-422
Chicken Lysozyme Type VI (from Egg White), MP Biomedicals
Enzymes
Lysozyme (muramidase) hydrolyzes preferentially the b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in the mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus.

  • Grade : Molecular Biology Reagent
  • Specific Activity : ≥23500 Shugar units/mg
  • Key Applications : Cell Lysis
  • Product Type : Proteins, Enzymes & Peptides
  • Biochemical Category : Proteins, Enzymes & Peptides
  • Extinction Coefficient : (E1%) 26.4 (280 nm) (Lit.)
  • Presentation : White to Off-white Powder
  • Format Type : VI, 3X Crystallized, Salt-Free, Albumin-Free, Lyophilized
  • Isoelectric point : (pI) pH 11.0 (lit)
  • pH : 3.6 (Lit)
  • NOTES : Chloride ≤3.5%

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty