Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate monohydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: Sodium Phosphate Monobasic hydrateSodium dihydrogen orthophosphate monohydrateSodium dihydrogenorthophosphate monohydrateSodium phosphate monobasic monohydrateSodium dihydrogen orthophosphate 1-hydrateSodium orthophosphate monobasic monohydrateSodium dihydrogen phosphate 1-hydrate

0219920201 02199202.5 0219920205 02199202.1
76177-500EA 144.32 USD InStock
76177-500 76177-498 76177-504 76177-502
Sodium dihydrogen phosphate monohydrate
Sodium dihydrogen phosphate monohydrate
Sodium phosphate is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography. Sodium phosphate occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Most neutral sodium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.
Sodium phosphate monobasic, Monohydrate is used as sequestrant, emulsifier and buffer in foods; as mordant in dyeing; for weighting silk in tanning; in manufacture of enamels, ceramics, detergents, boiler compounds; as fireproofing agent; in soldering and brazing instead of borax; as reagent and buffer in analytical chemistry; cathartic; laxative. A table for preparation of 0.1 M sodium phosphate buffer at 25 °C using various proportions of sodium phosphate monobasic and sodium phosphate dibasic has been published. A study of the effect of freeze-thaw storage cycles on proteins in sodium phosphate and potassium phosphate buffer solutions has been reported. The effect of 5 mM sodium phosphate on the efficacy of electrospray ionization (ESI) ion mobility spectrometry (IMS) analysis has been evaluated. A protocol for the purification of pyrogen-free mouse IgG1 monoclonal antibodies which uses 10 mM sodium phosphate (pH 7.4) has been published. An ion-pairing HPLC method for the analysis of 5-aminosalicylic acid has been reported. A TLC method for separation of nucleotide sugars in the study of glycosyltransferase activity has been published.
Room Temperature, Desiccate

  • USP Grade
  • Presentation: White Powder
  • Soluble in water (100 mg/mL), yielding a clear, colorless solution.


Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 214 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg
Order Now
Specification Test Results
Identity Test Passes
Assay 98.0-103.0%
pH (5% aq soln) 4.1-4.5
Water 10-15%
Insoluble Substances ≤0.2%
Chloride ≤0.014%
Sulfate ≤0.15%
Aluminum, Calcium & Related Elements Passes
Arsenic ≤8 ppm
Heavy Metals ≤0.002%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty