Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride ≥99.5%, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: HaliteSea saltSaline

0219473805 02194738.5 0219473810 0219473801
76177-746EA 63.97 USD InStock
76177-746 76177-744 76177-748 76177-750
Sodium chloride ≥99.5%, white powder cell culture reagent
Sodium chloride
Storage: Store at Room Temperature (15-30 °C)
Sodium chloride is a commonly used chemical which is found widely in nature. It is considered to be an essential nutrient. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.
Sodium chloride is used in a wide variety of biochemical applications, including intravenous fluids (normal saline is 0.9% w/v in water), density gradients, a diluent to increase ionic strength in buffers or culture media and in salt-out procedures in the isolation of DNA. It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions. Sodium chloride is widely used in protein crystallization.

  • Cell Culture Reagent Grade
  • Vapor Pressure: 1 mm Hg at 865 °C (lit.)
  • Presentation: White Powder
  • Soluble in water (357 mg/mL at 25 °C; 384 mg/mL at 100 °C), glycerol (100 mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl.


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C (1013 hPa) (lit.)
Melting Pt: 801 °C (lit.)
Density: 2.17 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/76/36400876.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99.5%
pH (5% aq soln) 5.0-9.0

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty