Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine ≥99%, white crystalline powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acidL(-)-MethionineMethionine(S)-(-)-2-Amino-4-(methylthio)butanoic acidH-Met-OH(S)-(-)-Methionine(S)-2-Amino-4-(methylthio)butanoic acid

0219470780 0219470725 0219470791
76177-872EA 623.73 USD InStock
76177-872 76177-868 76177-914
L-Methionine ≥99%, white crystalline powder cell culture reagent
L-Methionine
Storage: Store at Room Temperature (15-30 °C).
Methionine is one of the common sulfur-containing amino acids. Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.

  • Cell Culture Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: 2.28, 9.21 (Lit.)
  • Soluble at 2.5 g to 100 mL of water.


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: 280…282 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/62/36400862.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (1% aq soln) 5.6-6.1
Loss on Drying ≤0.3%
Optical Rotation (c=2, 6N HCl) +22.4° to 24.7°
Residue on Ignition ≤0.4%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty