Would you like to log in again?
PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Supplier: Gilson
PIPETMAN®
F171201 F171601 F171600 F171301 F171200 F171100 F171401 F171300 F171400 F161230 F171101 F161370 F171501 F171500
76178-190PK 120.8 USD InStock
76178-190 76178-208 76178-200 76178-202 76178-204 76178-206 76178-192 76178-184 76178-194 76178-186 76178-196 76178-210 76178-188 76178-198
PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Pipette Tips
When presented in convenient, steady, Tipack™ racks, Diamond Tips are easy to pick up directly from the rack.
  • To help prevent contamination, the 96 tips are loaded without manual intervention
  • Autoclaving conditions: Moist heat/121 °C/20 min/1 bar
  • Fully autoclavable and sealed with a security sticker
  • Color-coded to differentiate tip models
  • Indicate the compatible Gilson® pipettor models
Tipack™ racks are sealed with a security sticker, fully autoclavable, and they will resist being autoclaved several times. Using Gilson PIPETMAN® Tips is the only way to ensure accuracy and reproducibility across the full volume range of PIPETMAN® pipettors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty