Would you like to log in again?
PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Supplier: Gilson
PIPETMAN®
F171103 F171603 F171403 F171303 F171503 F171703 F171203 F171803
76178-212PK 231.71 USD InStock
76178-212 76178-222 76178-214 76178-224 76178-216 76178-226 76178-218 76178-220
PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Pipette Tips
Highest quality Diamond Tips for superior results.
Tipack™ racks are supplied as 10 packs of 96 tips
Autoclaving conditions: Moist heat/121 °C/20 min/1 bar. Remove blister packaging before autoclaving.
These PIPETMAN® Filter Tips play a critical role in preventing aerosol-borne contamination. High-density polyethylene filter stops particles down to 1 µm, providing an effective barrier against aerosols. PIPETMAN® Filter Tips are designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology, and cell technology. Sterilized filter tips are certified free of detectable RNase, DNase, DNA, RNA, and protease.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty