Would you like to log in again?
DistriTip® Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Supplier: Gilson
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Distritips®
F164100 F164110 F164120
76178-250PK 105.89 USD InStock
76178-250 76178-252 76178-254
DistriTip® Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Pipette Tips
The best quality to guarantee repetitive distribution results
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 1.25 ml. More than 100 aliquots can be dispensed from each syringe. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty