Would you like to log in again?
DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Supplier: Gilson
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Distritips®
F164150 F164140 F164130
76178-256PK 154.15 USD InStock
76178-256 76178-258 76178-260
DistriTip® Sterilized Syringe Tips for DISTRIMAN® Repetitive Pipettor, Gilson®
Pipette Tips
The best quality to guarantee repetitive distribution results.
DistriTip® tips are polypropylene syringes with easy-to-read level indicators. Three models cover the entire volume range from 1 µl to 12.5 ml. More than 100 aliquots can be dispensed from each syringe. These sterilized syringe tips are individually wrapped and certified free of RNase, DNase, DNA, RNA, and protease. Used with the DISTRIMAN® positive-displacement repetitive pipettor, DistriTip® tips provide more accurate and precise measurements for viscous, dense, volatile, or hot liquids.
DistriTip® Syringe Tips are specifically designed for use with the DISTRIMAN® Repetitive Pipettor.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty