Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium acetate 7.5 M, clear solution

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Acetic acid ammonium salt

0219875980 0219875991
76180-704EA 249.33 USD InStock
76180-704 76180-702
Ammonium acetate 7.5 M, clear solution
Ammonium acetate
Ammonium acetate is a chemical compound which is derived from the reaction of ammonia and acetic acid. Ammonium acetate is volatile at low pressures. Because of which it has been used to replace cell buffers with non-volatile salts, in preparing samples for mass spectrometry. It is used as a buffer for mobile phases for HPLC with ELSD detection.

Ammonium acetate is a widely used reagent in molecular biology and chromatography. Its applications include the purification and precipitation of DNA and protein crystallization. Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides. Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.

Store at +4 °C.

Presentation: Clear Solution


Formula: H₃CCOONH₄
MW: 77.08 g/mol
Density: 1.17 (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013066
CAS Number: 631-61-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg
Order Now
Specification Test Results
Identity Test Passes
Concentration 7.5 M aq (578.25 g/L)
Sterility Sterile

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty