Would you like to log in again?
FULL KIT MARVELX PEEK/316 SS 125UMX70MM
Supplier: IDEX HEALTH & SCIENCE LLC
Marvelx* Full Kit 125umx70mm^ Marvelx* Full Kit, Wetted Material: PEEK lined and 316 Stainless Steel, Installation Method: Fingertight 1/8 to1/4 of a turn after the initial resistance, Fitting Type: 10-32 threaded, Presure: 19,000 psi, Temperature: 120 deg C, Tubing Length: 70mm, Tubing ID: 125um
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FULL KIT MARVELX PEEK/316 SS 125UMX70MM 76181-078 UPFS-6125070 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty