Would you like to log in again?
Vials, National Scientific™, Thermo Scientific
Supplier: Thermo Scientific
MSCERT5000-S34W CERT5000-993 CERT5000-175W MSCERT5000-40W MSCERT5000-32 MSCERT5000-34W CERT5000-80W CERT5000-82W CERT5000-992 CERT5000-93W CERT5000-79 WSKT-25 CERT5000-78W CERT5000-92W CERT5000-176W CERT5000-580W CERT5000-593W CERT5000-69LV CERT5000-75W CERT5000-578W MSCERT5000-38 MSCERT5000-401W MSCERT5000-41W CERT5000-72LV CERT5000-80 CERT5000-193W
10222-192PK 230.62 USD InStock
10222-192 10222-194 10222-230 10222-274 10770-254 10222-238 10222-218 10770-290 10222-208 10754-522 10222-190 10770-306 10770-268 10222-240 10222-264 10754-526 10754-524 76045-292 10770-288 10222-226 10222-206 10770-280 75852-612 75852-614 10769-144
Vials, National Scientific™, Thermo Scientific
Vials
These vials are fully lot tested including HPLC and GC analysis for 15 critical parameters.
  • Superior quality 1st hydrolytic class glass (Type 1) in clear and amber
  • Fixed insert or conical base vials for limited volume sampling
Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass.
Delivery information: Vials are available in assembled and unassembled kits.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty