Would you like to log in again?
Chromacol Headspace Caps and Septa
Supplier: Thermo Scientific
WSKTCL-100
76181-356PK 78.26 USD InStock
76181-356
Chromacol Headspace Caps and Septa
Closures Chromatography Vial Closures
Chromacol headspace caps and septa use magnetic seals with PAL/CTC/Leap Technologies, Gerstel and other magnetic transport autosamplers.
  • 20 mm crimp seals must be applied with a crimping tool
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Extended temperature range products for high-temperature headspace applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty