Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BriskHeat® Silicone-Rubber Heater Blankets with Control and Adhesive, for Metal Surfaces, 120VAC, Cole-Parmer

Supplier: Cole Parmer

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BriskHeat®
36002-21 36002-25 36002-01 36002-13 36002-05 36002-09 36002-29 36002-17 36001-98
76181-618EA 1020.87 USD InStock
76181-618 76181-606 76181-616 76181-604 76181-608 76181-610 76181-620 76181-614 76181-612
BriskHeat® Silicone-Rubber Heater Blankets with Control and Adhesive, for Metal Surfaces, 120VAC, Cole-Parmer
Heaters Heating Mantles
Rugged, moisture/chemical resistant heaters provide uniform heat distribution across large surfaces.

  • Maximum exposure temperature of 425 °F (218 °C)
  • Minimum temperature of 50 °F (10 °C)
  • Lead length 72"
  • US Standard plug type
  • Easy-to-apply industrial strength pressure sensitive adhesive backing

These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.

Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 425°F. No additional control is necessary.

The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 W/in² power density.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty