Would you like to log in again?
Cole-Parmer® XRF Window Films
Supplier: SPEX SamplePrep
Cole-Parmer®
3520 3517 3511 3525 3516
76181-868RL 112.06 USD InStock
76181-868 76182-372 76182-374 76181-872 76182-366
Cole-Parmer® XRF Window Films
Sample Containers XRF Sample Cups
Thin window film for XRF liquid sample cups
  • Compatible with X-Cell® sample cups
Cole-Parmer® XRF window film serves as the sample window over our X-Cell sample cups, allowing for uniform transmission of x-rays while effectively containing the liquid sample.
Kapton® polyimide film is chemically imperious to almost anything except strong alkali solutions. It is often considered the closest thing to a bombproof XRF window film due to its incredible strength and heat resistance to over 300°C (572° F). The phosphate-based surface coating improves its handling characteristics.
BoPET (Mylar®) film is unusually strong for its thickness, it is resistant to organic chemicals but vulnerable to strong acids and bases. It may contain trace levels (ppm) of Ca, P, Fe, Cu, Zn, or Sb.
Polypropylene film is reasonably resistant to acids, bases, and many organic chemicals, but can be attacked by aromatic and halogenated hydrocarbons. It may contain trace levels (ppm) of Ca, Zr, P, Fe, Zn, Cu, Ti, and Al.
Ultralene® film offers the best balance between X-ray transmission, chemical resistivity, strength, and purity. The 4 µm (0.16 mil) thickness allows for a greater degree of X-ray transmission and makes it ideal for light-element analysis. It is heat resistant up to 100°C (212°F) and is reasonably resistant to acids, bases, and many organic chemicals, but can be attacked by aromatic and halogenated hydrocarbons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty