Would you like to log in again?
Cole-Parmer® XRF Spec-Caps® Cups
Supplier: SPEX SamplePrep
Cole-Parmer® XRF Spec-Caps®
3619 3617 3615 3619AM 3615M 3617M 3619M 3619A 3618
76181-904BG 458.28 USD InStock
76181-904 76181-926 76181-914 76181-902 76181-924 76181-918 76181-916 76181-922 76181-920
Cole-Parmer® XRF Spec-Caps® Cups
Mills
Aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
  • Reinforces sample disks to resist chipping and breaking
  • Makes sample disks easier to remove from the pellet die
  • Minimizes contamination during sample handling
  • Protects the bore of the pellet die from abrasive samples
Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets. Preflared cups come with an unpainted surface for a snug fit in the pellet die. Unflared cups feature a pained outer surface to prevent seizure in the die and allow for labeling. Edge flaring prevents sample loss and pellet jamming. Utilize the edge flaring tool to help create a tighter fit in the pellet die. To use, simply press the tapered end of the edge flaring tool into the Spec-Caps® cup.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty