Would you like to log in again?
Cole-Parmer® Graphite Crucibles
Supplier: SPEX SamplePrep
Cole-Parmer®
7159 7152HP 7157 7158 7161
76182-060PK 171.51 USD InStock
76182-060 76182-240 76182-460 76182-456 76182-458
Cole-Parmer® Graphite Crucibles
Crucibles
Cost-effective alternative to precious metal crucibles.
  • Chemically inert and heat resistant
  • Disposable
Disposable crucibles eliminate the need for time-consuming cleaning procedures and the possibility of cross-sample contamination. Regular-purity crucibles are made of graphite with impurities ranging from 10 to 100 ppm each of Mg, Fe, Al, Si, and Cu. They are well suited for most basic borate fusion applications.
High-purity crucibles are made of graphite with total metallic impurity limits of 2 ppm—ideal for trace-level analytical work.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty