Would you like to log in again?
Cole-Parmer® SM-200 ShatterBox® Versatile Ring and Puck Mill
Supplier: SPEX SamplePrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cole-Parmer®
8507 8504 8530-115 8505 31688 8507R 8501 8521 51793 8508 8506
76182-526EA 16291.09 USD InStock
76182-526 76181-946 76182-528 76182-530 76182-364 76182-532 76182-114 76182-116 76182-534 76182-536 76182-524
Cole-Parmer® SM-200 ShatterBox® Versatile Ring and Puck Mill
Mills
Versatile Ring and Puck Mill with soundproof enclosure keeps noise level down while grinding samples.
  • Grind samples up to 150 g to analytical powder in minutes
  • Dish-shaped grinding container swings in a tight high-speed circle to crush samples
  • Membrane control panel features an LCD with 10-minute electronic timer
  • Lockable caster wheels ensure mill stays in place during operation
SM-200 ShatterBox® versatile ring and puck mill is ideal for pulverizing dry, brittle samples and slurry grinding. To use, place your sample between the puck and the wall of the grinding container, place the clamp over the container, then simply set the timer and run the mill. A puck and ring swing around inside a grinding container at a high speed to crush the samples to analytical quality powder.
Clamp adjusts to fit various grinding containers. Safety interlock system and lid gas cylinders offer operator safety. A 10-minute timer counts down the time remaining. Press 'Pause' any time to interrupt the grinding cycle. Grinding containers are available in various sizes and materials.
Typical samples include include cements, soils, ceramics, slag, rocks, minerals, ores, pharmaceuticals, sulfur pellets, and dried plant material.
Ordering information: Order rack 8507R to hold one or three small grinding containers (8507 and 8508) in the chamber.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty