Would you like to log in again?
Cole-Parmer® Freezer/Mill® Cryogenic Grinder Accessories
Supplier: SPEX SamplePrep
Cole-Parmer®
6870M-1 6870S 6756 6758 6755 6791 6754 6906 6884 6807 6805 6808M 6887 6905 6870L 6808 6804 6888 6907
76181-958EA 1075.43 USD InStock
76181-958 76182-440 76182-044 76182-450 76182-032 76182-438 76182-046 76182-442 76182-430 76182-034 76182-444 76182-048 76182-234 76182-432 76182-446 76182-226 76182-434 76182-228 76182-436
Cole-Parmer® Freezer/Mill® Cryogenic Grinder Accessories
Mills
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes.
Compatible with Cole-Parmer® Freezer/Mill® cryogenic grinders 6775 and 6875.
These accessories are ideal for optimizing performance of grinding tissue, seeds, polymers, pharmaceuticals, and textiles. The dewar and cryogenic hoses are ideal for manually dispensing liquid nitrogen directly into the Freezer/Mill® cryogenic grinders. The end plug extractors easily remove end plugs from frozen vials. Simply place a vial into the extractor and press down on the handle. Samples can be removed from the vial while cold. Suction cups on the extractor offer secure benchtop placement for one-handed operation. Vial extractors lift grinding vials in and out of the Freezer/Mill® cryogenic grinders. Vial extractors assist with easy retrieval of impactors from vials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty