Would you like to log in again?
Cole-Parmer® Deuterium Lamps
Supplier: Antylia Scientific
Cole-Parmer®
1194116 11941-26 11941-27 11941-31 11941-11 11941-34 11941-33 11941-25 11941-17 11941-29 11941-30 11941-28 11941-15 11958-00 11941-12 11941-16 1194124 11941-35 11941-21 11941-59 1194123 11941-32 11941-14 1194111 11941-13 11941-58 11941-19 11941-20 11941-24 11941-36 11941-10
76473-760EA 1107.27 USD InStock
76473-760 76473-784 76473-762 76182-490 76182-494 CA76181-236 CA76181-238 76473-764 76473-744 76182-616 76182-618 76182-496 76182-510 76182-498 76182-480 76182-482 76182-484 CA76181-242 76473-754 76182-626 76182-504 CA76181-240 76182-628 76182-506 76182-508 76182-486 76182-488 76182-620 76182-622 76182-624 76182-502
Cole-Parmer® Deuterium Lamps
Lights Deuterium Lamps
Economical replacement deuterium lamps.
  • Select from standard or long life lamps
  • Manufactured in an ISO accredited facility
  • Individually tested and issued with a Test Certificate
This range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. The lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
These deuterium lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty