Would you like to log in again?
TORCH HAND CUTTING 90DEG HEAD 20-1/2 IN
Supplier: ORS NASCO, INC.
Heavy Duty Hand Cutting Torch, 90A Head, Oxygen/Fuel Gas^ Heavy Duty Hand Cutting Torch, 90A Head,Oxygen/Fuel Gas, Cuts Up to 12in,20 1/2 L, Body Material: BRASS, SST, Connections: 9/16 - 18, Cutting Cap. [Max] 12 in, Gas Type: All; Oxygen/Fuel Gas, Lengt
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TORCH HAND CUTTING 90DEG HEAD 20-1/2 IN 76183-070 635-SC229 Each (21Inch) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty