Would you like to log in again?
TORCH CUTTING GAS AXE 90 D HV DUTY 21IN
Supplier: ORS NASCO, INC.
Gas Axe Extra Heavy Duty Cutting Torch^ Gas Axe, Extra Heavy Duty Cutting Torch, Cutting Cap. [Max] 24 in, Cutting Cap. [Min] 20 in, Gas Type Acetylene; Propylene, Head Angle 90 deg, Length [Nom] 21 in, Type Extra Heavy Duty Cutting Torch, Used on Torch H
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TORCH CUTTING GAS AXE 90 D HV DUTY 21IN 76183-074 635-SC929 Each (21Inch) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty