Would you like to log in again?
CUTTING TIP HEAVY-DUTY SC12 SERIES SZ3
Supplier: ORS NASCO, INC.
SC12 Series Heavy-Duty Cutting Tips, Size 3, 1 1/2 in - 2 in Thick^ SC12 Series Heavy-Duty Cutting Tips, Size 3, 1 1/2 in - 2 in Thick, Flow Range 180 cfh, Gas Type Acetylene; Oxygen/Acetylene, Head Type Concaved, Material: Metal, Thickness [Nom] 1 1/2 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CUTTING TIP HEAVY-DUTY SC12 SERIES SZ3 76183-090 635-SC12-3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty