Would you like to log in again?
DAVISIL® Grade 923 Chromatographic Silica, W.R. Grace
Supplier: W.R. Grace
5173174 5173172 5183338 5183337 5007039 5173173
76183-106EA 2478.09 USD InStock
76183-106 76183-104 76183-168 76183-102 76183-166 76183-108
DAVISIL® Grade 923 Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Grade 923 meets the specification for Silica in the ASTM D-1319 FIA test and the ASTM D-2549 Test.
  • Original silica specified in ASTM
  • Proven performance
  • Exceptional quality and consistency
Grace® Grade 923 Chromatographic Silica is a narrow pore type silica gel which meets the requirements for Silica Gel set forth in the procedure ASTM D 1319 and IP 156, as well as ASTM D 2549. Available in several convenient package configurations.
Certifications: Meets Specification for Silica in ASTM D-1319 and ASTM D-2549.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty