Would you like to log in again?
Kinesis® TELOS® neo™ MicroPlate™ SPE Loose Wells, Cole-Parmer
Supplier: Argos Technologies
06474-20 06474-08 06474-17 06474-01 06474-14 06474-12 06474-10 06474-04 06474-16
76183-268PK 565.77 USD InStock
76183-268 76183-254 76183-276 76183-286 76183-284 76183-272 76183-260 76183-292 76183-280
Kinesis® TELOS® neo™ MicroPlate™ SPE Loose Wells, Cole-Parmer
Microplates Solid Phase Extraction Plates (SPE)
Populate base plate with only the wells needed.
  • 50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
The internal diameter, frit design, and sorbent material of the TELOS MicroPlate loose wells allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Use the loose wells for assays when the sample numbers vary, populating the base plate with only the required number of wells. Base plates are required to use loose wells. Full or partially populated plates can be processed using either vacuum or positive pressure. Order method development kit to build a method development plate to suit your applicatiion.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty