Would you like to log in again?
Digi-Sense® Mini Vane Anemometer with NIST-Traceable Certificate, Cole-Parmer
Supplier: Cole Parmer
Digi-Sense®
20250-23
76183-374EA 131.31 USD InStock
76183-374
Digi-Sense® Mini Vane Anemometer with NIST-Traceable Certificate, Cole-Parmer
Anemometers
Verify proper airflow for component or space cooling with lightweight and pocket-sized affordable meter for convenient field use.
  • Reliable measurements guaranteed
  • Pre calibrated to NIST-traceable standards
  • User-selectable units of measure (m/s, k/hr, ft./min., mph, or knots) eliminate the need for conversion
  • Measure air velocities up to 30.0 m/s; 5900 fpm
  • Max./Avg. functions allow easy management of airflow measurement
  • Unique backlight LCD sensor is automatically activated by low-light conditions; no need to manually adjust
  • Auto power-off after 15 minutes of non-use extends battery life
  • Low-battery indicator prompts battery replacement and prevents power loss
The Digi-Sense compact vane anemometer is designed for precision airflow measurements in five user-selectable units of measure (ft/min, m/s, mph, km/h, or knots). The instrument will help you meet cooling and environmental requirements for safety and comfort. Small size, comfort-grip form, and ergonomically placed push buttons allow you to easily select either maximum or average air velocity readings. Auto power-off after 15 minutes of nonuse extends battery life. Low-battery indicator prompts battery replacement, preventing power loss.
Digi-Sense comes pre-calibrated, saving you extra waiting time and costs to calibrate separately.
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery information: Includes one 9V battery (100 hours typical) and NIST-traceable calibration report with data supplied by InnoCal.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty