Would you like to log in again?
CELLvo™ XF Matrix, StemBioSys
Supplier: StemBioSys
XF-HPME-6WP XF-HPME-T75 XF-HPME-T150
76183-424PK 422.86 USD InStock
76183-424 76183-422 76183-420
CELLvo™ XF Matrix, StemBioSys
Cell Culture Supplements and Additives
CELLvo™ XF Matrix is a ready-to-use cell culture substrate which provides a xeno-free native microenvironment that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate.
  • CELLvo™ XF Matrix is a natural extracellular matrix synthesized in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow
  • CELLvo™ XF Matrix is an advanced stem cell culture system that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix proliferate more rapidly
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix maintain a smaller cell size
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ XF Matrix is a protein extracellular matrix produced under xeno-free conditions that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
CELLvo™ XF Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes.
Accessories information: Accessories not included.
Delivery information: CELLvo™ XF Matrix arrives at room temperature but should be refrigerated at 2 to 8 °C upon arrival. Avoid extended exposure to light.
Packaging: CELLvo™ XF Matrix is produced under aseptic conditions and should be handled using aseptic techniques.
Caution: Handle CELLvo™ related products in accordance with established bio-safety practices.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty