Would you like to log in again?
Digi-Sense® Thermocouple General-Purpose Air/Gas Probes, Cole-Parmer
Supplier: Cole Parmer
08516-75 08500-75 08517-75
76183-446EA 102.55 USD InStock
76183-446 76183-356 76183-590
Digi-Sense® Thermocouple General-Purpose Air/Gas Probes, Cole-Parmer
Temperature Probes
Air/Gas Probes are ideal for measuring a wide range of air temperatures.
  • Response time: 1.75 seconds or greater
  • Exposed Junction is protected by a rigid stainless-steel shield minimize error from radiated heat
  • Each probe comes with a color-coded ANSI coiled cable: Type J-black, Type K-yellow, Type T-blue, and Type E-purple
  • Custom designs or changes to this product are available upon request
Temperature accuracy for the Type–T probe is ±3.6 °F (±2.0 °C), or ±0.4% of reading above 32 °F (0 °C); ±0.8% of reading below 32 °F (0 °C). Temperature accuracy for the Type–K probe is ±3.0 °F (1.1 °C) or ±0.4% reading above 212 °F(100 °C); ±3.6 °F (2.2 °C) or 2% of reading between –58 to 211 °F (–50 to 99 °C); ±7.2 °F (4 °C) or ±4% of reading between –60 to –238 (–51 to –150 °C), whichever is greater. Temperature accuracy for the Type–J probe is ±5.9 °F (±3.3 °C), or ±0.4% of reading above 32 °F (0 °C); ±4.0% of reading below 32 °F (0 °C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty