Would you like to log in again?
Digi-Sense® Benchtop Temperature Limit Controllers, Cole-Parmer
Supplier: Cole Parmer
36225-77 36225-79 36225-78 36225-76
76183-608EA 1509.14 USD InStock
76183-608 76183-606 76183-604 76183-602
Digi-Sense® Benchtop Temperature Limit Controllers, Cole-Parmer
Controllers Temperature Controllers
A secondary safety device that cuts power when a setpoint is achieved.
  • Models available for type J, K, or T thermocouples, or RTD probes
  • Emergency power cutoff with audible alarm protects processes from dangerous overshoot
  • Manual reset required after alarm condition to enable power up
Ordering information: Includes 6 ft. detachable power cord and lattice support bracket.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty