Would you like to log in again?
Digi-Sense® Benchtop PID Temperature Controllers, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
36225-64 36225-62 36225-63 36225-61
76183-612EA 1241.66 USD InStock
76183-612 76183-610 76183-616 76183-614
Digi-Sense® Benchtop PID Temperature Controllers, Cole-Parmer
Controllers Temperature Controllers
Perfect for basic temperature control applications.
  • Models available for type J, K, or T thermocouples, or RTD probes
  • Displays both setpoint and current temperature for convenience
  • Auto tune functions to stabilize temperature at set point and minimize overshoot
  • Audible alarm alerts user to unsafe temperature conditions
Ordering information: Includes 6 ft. detachable power cord and a lattice support bracket.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty