Would you like to log in again?
Esteem® TRU-BLU™ Nitrile Examination Gloves, Cardinal
Supplier: Cardinal
Esteem® TRU-BLU™
8897NB 8899N 8899NB 8898NB 8899NXX 8899NXXB 8896NB 8896NB 8895N 8895NB
76183-724CS 678.74 USD InStock
76183-724 CA89040-196 76183-726 CA89040-206 76183-728 76183-718 CA89040-204 CA89040-198 76183-720 76183-722
Esteem® TRU-BLU™ Nitrile Examination Gloves, Cardinal
Gloves
Stretchy, powder-free nitrile gloves feature a high coefficient of friction (grip), even when wet
  • Beaded cuff resists tearing and minimizes cuff rolldown
  • Microtextured for excellent grip in wet or dry applications
  • Ambidextrous
  • Thickness: 5.1 mil (finger)
They also provide good tactile sensitivity while maintaining film strength and barrier protection. Gloves have been tested for resistance to chemotherapy agents and for use with common hospital and laboratory chemicals.
Caution: Note: Gloves used for protection against chemotherapy drug exposure must be selected specifically for the type of drugs being used.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty