Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
High Density Twist Knot Wire Wheel, Weiler®
Supplier: Weiler
804-09799 804-08309
76183-772CT 557.65 USD InStock
76183-772 76183-762
High Density Twist Knot Wire Wheel, Weiler®
Abrasives
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
  • Extra high density for superior performance and high aggression
  • Heavy-duty brushing action with some flexibility
  • Steel filter material
Use for cleaning, heavy burr removal, roughening surfaces, and surface conditioning applications. Removes rust, paint, oxides, and scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty