Would you like to log in again?
TIGER® X, Coated Abrasive Flap Discs, Weiler®
Supplier: Weiler
804-51229 804-51209 804-51222 804-51233 804-51228 804-51201 804-51207 804-51226 804-51227 804-51223 804-51206 804-51200 804-51214 804-51234 804-51213 804-51210 804-51212 804-51230 804-51216 804-51205 804-51202 804-51218 804-51219 804-51204 804-51232 804-51221 804-51211 804-51208 804-51217 804-51224 804-51215 804-51225 804-51203 804-51220 804-51231
76183-938PK 139.55 USD InStock
76183-938 76184-042 76184-064 76184-020 76184-062 76184-040 76184-060 76184-026 76184-028 76184-048 76184-022 76184-046 76184-024 76184-066 76184-044 76184-008 76184-052 76184-050 76184-030 76183-940 76183-942 76184-116 76184-014 76184-036 76184-058 76184-016 76184-038 76184-056 76184-010 76184-032 76184-054 76184-034 76184-012 76184-018 76184-118
TIGER® X, Coated Abrasive Flap Discs, Weiler®
Abrasives
Features advanced grain anchoring system
  • Phenolic backed body material
  • Dark gray finish
  • Use with right angle grinders
Use for weld blending, flat surface grinding, finishing, heavy stock removal, light pressure blending, and deburring applications.
Ideal for use on steel, stainless steel, cast iron, and non-ferrous soft metals. Delivers faster grinding and longer life when used on steel and stainless steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty