Would you like to log in again?
Tiger, Ceramic Angled Flap Discs, Weiler®
Supplier: Weiler
Tiger®
804-51316 804-51314 804-51313 804-51312 804-51315 804-51317
76184-098BX 206.49 USD InStock
76184-098 76184-088 76184-096 76184-094 76184-092 76184-090
Tiger, Ceramic Angled Flap Discs, Weiler®
Abrasives
Deliver an aggressive, long life solution for grinding fillet welds, T-joints and other hard-to-reach places.
  • Aggressive grinding; attacks metal, removing more material in less time for faster results
  • While grinding fillet welds, the angled flaps reduce the risk of gouging the work-piece
  • Angled flaps allows grinding at a higher or lower angle, getting into hard-to-reach places more easily
The 90° angled flap design forms perfectly into angles while the phenolic back provides stability to the flaps. This combination optimizes flap-to-metal contact, resulting in exceptional feel and control for the operator while grinding and finishing. Top-sized Ceramic Alumina grain offers a contaminant-free option for stainless steel applications. Grinds cool, delivering maximum performance on Inconel, titanium and other hard-to-grind metals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty