Would you like to log in again?
FLAP DISC COATED TIGER PAW 5X0.35X5IN
Supplier: ORS NASCO, INC.
Tiger Paw* Coated Abrasive Flap Disc 5 in^ Tiger Paw* Coated Abrasive Flap Disc, Designed for aggressive performance on tough grinding applications, High performance grain and special poly cotton cloth provide superior grain retention, Height: 5 in, Length: 0.35 in, Width: 5 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLAP DISC COATED TIGER PAW 5X0.35X5IN 76184-108 804-51135 Carton of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty