Would you like to log in again?
Tiger, Ceramic Combo Wheels, Weiler®
Supplier: Weiler
Tiger®
804-58315 804-58316
76184-234BX 164.06 USD InStock
76184-234 76184-232
Tiger, Ceramic Combo Wheels, Weiler®
Abrasives
Self-sharpening ceramic alumina grain delivers a superior combination of life and cut rate
  • Superior life and cut on metal
Use for cutting, edge beveling, edge softening, grinding, notching, scale removal, stock removal, weld blending, and weld removal applications.
Ideal for stainless steel and other hard-to-grind metals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty