Would you like to log in again?
Tiger, Ceramic Resin Fiber Discs, Weiler®
Supplier: Weiler
Tiger®
804-69884 804-69852 804-69880 804-69883 804-69882 804-69851 804-69854 804-69881 804-69850 804-69853
76184-632BX 70.3 USD InStock
76184-632 76184-666 76184-630 76184-664 76184-672 76184-670 76184-628 76184-626 76184-668 76184-624
Tiger, Ceramic Resin Fiber Discs, Weiler®
Abrasives
Advanced ceramic grain leverages a crystalline structure that is significantly finer than standard ceramic
  • Advanced ceramic grain delivers greater stock removal with a finer scratch pattern
  • Less pressure required to achieve consistent material removal rate
  • Use with right angle grinders
  • Resin bonded
Advanced ceramic grain removes material effortlessly while a top coat grinds cool and eliminates loading and heat buildup. For those that put a premium value on getting work done faster and optimizing disc usage, Tiger Ceramic delivers.
Additional fracture points result in a more aggressive grain that self sharpens at a higher rate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty