Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Dually™ Stringer Bead Wheel, Weiler
Supplier: Weiler
804-79802 804-79805 804-79801 804-79800 804-79804
76184-720EA 48.54 USD InStock
76184-720 76184-728 76184-726 76184-724 76184-722
Dually™ Stringer Bead Wheel, Weiler
Abrasives
Up to the task for the most demanding multi-pass welding applications.
  • 0.02" carbon steel bristle
  • One-piece dual hex-nut that eliminates interference issues and simplifies periodic flipping of the wheel
  • Delivers fast and effective cleaning action
  • Long life, safe use
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty