Would you like to log in again?
Total Collagen Assay Kit (Perchlorate-Free), Biovision
Supplier: BioVision
AB222942-100TEST
76186-154EA 1114.28 USD InStock
76186-154
Total Collagen Assay Kit (Perchlorate-Free), Biovision
Assays Protein Assays
A simple, sensitive & easy colorimetric assay to quantitate total collagen levels.
  • Simple, fast and convenient assay
  • Can detect as low as 0.5 µg of collagen in a variety of samples
  • Simple procedure; takes less than ~60 minutes
  • Faster than Perchlorate-Based Assay
  • PCA-free
Collagen is the most abundant protein in mammals, accounting for nearly 30% of body protein content. Collagen is an extracellular scaffolding protein found in skin, tendons, bone, cartilage, ligaments and muscles, amongst other tissues. There are more than twenty different characterized types of collagen, but the fibrilous types I, II and III comprise more than 90% of the total collagen in mammals. Collagen has a triple helical structure, consisting of three ‘preprocollagen’ strands with a repeating Gly-X-Hyp tripeptide sequence, where X is any amino acid and Hyp is hydroxyproline, a post-translationally modified amino acid formed from proline by the enzyme prolyl-4-hydroxylase. In animals, hydroxyproline is found almost entirely in collagen (which contains approximately 12-14% hydroxyproline by mass) and contributes to its structural rigidity and high tensile strength. Hydroxyproline is thus a direct measure of the amount of collagen present in tissues. Many diseases are believed to affect collagen turnover, including tumor invasion and metastasis, rheumatoid arthritis, cardiopulmonary fibrosis and muscular dystrophy. BioVision’s Total Collagen Assay Kit II (Perchlorate-Free) is a quick and convenient protocol for detection of small amounts of collagen in a variety of biological samples. The assay is based upon hydrolysis of samples to yield free Hyp, which is oxidized and reacted to give a brightly-colored chromophore with an absorbance peak at 560 nm. The assay employs a proprietary acidic developer solution to accurately measure collagen in hydrolysates without the use of hazardous perchloric acid, obviating the need for any special handling and waste-disposal protocols. The assay can detect a minimum of 0.5 µg collagen per well in a 96-well format and generates colorimetric results that are virtually identical to those of the classical perchloric acid-based method
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty