Would you like to log in again?
DuPont™ Tychem® 5000 Encapsulated Level B Suits
Supplier: DuPont
Tychem®
C3528TTN3X000600 C3528TTNXL000600 C3528TTNMD000600 C3528TTNLG000600 C3528TTN2X000600 C3528TTN4X000600 C3528TTNSM000600
76187-420CS 1526.91 USD InStock
76187-420 76187-344 76187-422 76187-424 76187-426 76187-428 76187-418
DuPont™ Tychem® 5000 Encapsulated Level B Suits
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Flat back
  • Front entry
  • Standard, single-layered 20 mil PVC visor
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Airline access
  • One exhaust vent
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment. Suits also provide coverage from overhead liquid splash (does not provide vapor protection). Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Standard faceshield is made from 20 mil PVC. Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment. Expanded back to accommodate self-contained air breathing apparatus (SCBA) flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Elastic openings at wrists allow for a tighter fit. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty