Would you like to log in again?
DuPont™ Tychem® 5000 Jacket
Supplier: DuPont
Tychem®
C3670TTNLG0006JF C3670TTN2X0006JF C3670TTN3X000600 C3670TTNXL0006JF C3670TTNXL000600 C3670TTNMD000600 C3670TTN3X0006JF C3670TTN4X000600 C3670TTNSM000600 C3670TTN2X000600 C3670TTNLG000600 C3670TTNMD0006JF C3670TTN4X0006JF
76187-450CS 414.79 USD InStock
76187-450 76187-440 76187-452 76187-442 76187-454 76187-432 76187-444 76187-434 76187-456 76187-446 76187-436 76187-448 76187-438
DuPont™ Tychem® 5000 Jacket
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Mandarin collar
  • Elastic wrists or jam fit cuff
  • Extends to hip
  • Double storm flap with hook & loop closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These jackets are tan.
These jackets are ideal for applications that do not require the full body coverage of a coverall. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Mandarin collar is designed to fit tightly around hooded PAPR. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Elastic openings at wrists allows for a tighter fit. Also available with jam fit cuffs, which is a CPE sleeve cuff plus glove insert for customer-installed jam fit glove systems. These jackets are typically worn with the C3360T pants or sold as jacket/pant ensemble (C3750T).
Certifications: Thickness meets ASTM D1777.
Ordering information: This jacket is available with elastic wrists or a jam fit cuff.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty