Would you like to log in again?
DuPont™ ProShield® 50 Coveralls with Respirator Hood
Supplier: DuPont
ProShield®
NB127SWHMD002500 NB127SWH5X002500 NB127SWHXL002500 NB127SWHLG002500 NB127SWH2X002500 NB127SWHSM002500 NB127SWH4X002500 NB127SWH6X002500 NB127SWH3X002500
76187-480CS 130.28 USD InStock
76187-480 76187-482 76187-350 76187-352 76187-474 76187-476 76187-478 76187-346 76187-348
DuPont™ ProShield® 50 Coveralls with Respirator Hood
Disposable Garments
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap
  • Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include: janitorial, sanitation, and general industrial maintenance.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty