Would you like to log in again?
DuPont™ Tychem® 10000 FR Encapsulated Level A Suits, Certified to NPFA 1991
Supplier: DuPont
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tychem®
RF600TSVLG00 RF600TSV4X00017S RF600TSVLG00017N RF600TSVSM000100 RF600TSV4X000100 RF600TSV2X000100 RF600TSV3X00 RF600TSVMD000100 RF600TSV3X00017S RF600TSV2X00017N RF600TSVXL00017N RF600TSV3X00017N RF600TSVLG00017M RF600TSV3X00017M RF600TSVLG00017S RF600TSV4X00017M RF600TSV2X00017S RF600TSV3X000100 RF600TSVXL000100 RF600TSVXL00017S RF600TSVXL00017M RF600TSVSM00 RF600TSVMD00017M RF600TSVXL00 RF600TSV2X00 RF600TSV2X00017M RF600TSVLG000100 RF600TSVMD00 RF600TSV4X00017N
76187-590CS 5258.93 USD InStock
76187-590 CA13452-728 76187-592 76187-594 76187-596 76187-598 13452-730 76187-610 76187-612 76187-614 76187-616 76187-606 76187-608 CA13452-730 13452-728 13452-726 13452-724 76187-584 13452-722 76187-586 13452-720 76187-588 76187-600 76187-602 76187-604 CA13452-720 CA13452-722 CA13452-724 CA13452-726
DuPont™ Tychem® 10000 FR Encapsulated Level A Suits, Certified to NPFA 1991
Disposable Garments
DuPont™ Tychem® 10000 FR NFPA 1991 (2016 edition) certified suits were one of the first limited-use garments to be certified to NFPA 1991 (2016 edition) in a single-skin construction.
  • Certified to NPFA 1991
  • Expanded back
  • Front entry
  • Triple-layered visor
  • Attached dual-layer gloves
  • Overgloves included
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Two exhaust valves
  • Triple taped seams
Tychem® 10000 FR garments are composed of a patented, heavy-duty aluminized fabric made of a DuPont™ Nomex®/DuPont™ Kevlar® blend laminated to a multi-layer barrier film. This unique combination of layers provides excellent physical strength and cut and abrasion protection. Ideal for industrial chemical, HazMat response and military applications including chemical warfare agents. Patented design is certified compliant to the 2016 edition of NFPA 1991, Standard on Vapor-Protective Ensembles for Hazardous Materials Emergencies, without requiring overgarment. Compliant with Base Requirements and Chemical Flash Fire Protection Option and Liquefied Gas Protection Option Requirements of NFPA 1991, 2016 edition.
The encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Triple taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing. Faceshield is triple-layered (40 mil PVC/ 5 mil Teflon® FEP/ 40 mil PVC) and standard width. Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Coveralls feature a vapor tight zipper, which is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Three component glove system features inner North Silver Shield® / 4H® glove provides broad range chemical protection, middle Neoprene gloves provide mechanical and physical durability to protect inner SilverShield® glove and outer knit glove made of Kevlar® for abrasion and cut resistance. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion. Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Internal adjustable waist belt system provides extra support and improved fit. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Certifications: Thickness meets ASTM D1777. Certified to NFPA 1991 (2005 edition). Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant.
Ordering information: Passthroughs are optional. Coveralls available with either (p/t) MSA Dual Purpose #495670, (p/t) MSA Quick fill #80311, or (p/t) Scott® #803620-01 Hansen fitting on right side.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty