Would you like to log in again?
DuPont™ Tychem® 6000 Coveralls with Respirator Hood
Supplier: DuPont
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tychem®
TF145TGY6X000600 TF145TGY5X0006TV TF145TGYSM0006TV TF145TGYMD000600 TF145TGY4X000600 TF145TGYXL0006TV TF145TGY7X000600 TF145TGY5X000600 TF145TGYMD0006TV TF145TGYLG0006TV TF145TGY3X000600 TF145TGY2X000600 TF145TGYXL000600 TF145TGY2X0006TV TF145TGYLG000600 TF145TGYSM000600 TF145TGY3X0006TV TF145TGY4X0006TV
76187-650CS 419.81 USD InStock
76187-650 76187-640 76187-652 76187-630 76187-642 76187-620 76187-632 76187-644 76187-622 76187-634 76187-646 76187-624 76187-636 76187-648 76187-626 76187-638 76187-628 76187-618
DuPont™ Tychem® 6000 Coveralls with Respirator Hood
Disposable Garments
Tychem® 6000 garments are made of a proprietary barrier film laminated to a Tyvek® substrate.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap
  • Taped seams
The fabric provides at least 30 minutes of protection against over 181 challenge chemicals including chemical warfare agents and toxic industrial chemicals. Tychem® 6000 garments are strong, durable and lightweight, and they are available in both high-visibility orange and low-visibility gray fabrics, making them a preferred choice for law enforcement, emergency medical services (EMS) technicians and military personnel.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Three-piece hood design with longer zipper that extends to chin allows for complete coverage of neck area and good fit around a respirator. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists and ankles allow for a tighter fit.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty